Learn More
MSC E.R. Wagner Stem Mount Swivel Caster, 4 in. Diameter, Polyurethane, 275 lb Capacity
Catalog No. 5000534159
Encompass
E.R. Wagner stem mount swivel casters are designed for industrial and commercial applications. This caster features a 4 in. diameter polyurethane wheel, a 1/2-13 threaded stem, and a 275 lb capacity, ensuring durable performance and reliable mobility.
  • Wheel diameter is 4 in.
  • Wheel material is polyurethane and rubber
  • Caster type is stem mount swivel
  • Capacity is 275 lb
  • Bearing type is Delrin
  • Stem type is threaded
  • Stem diameter is 1/2-13
  • Construction is hardened-heat treated
  • Application for industrial and commercial use
  • Finish is zinc chromate
Catalog No. 50-005-34159 Supplier MSC Supplier No. 01506062
E.R. Wagner stem mount swivel casters are designed for industrial and commercial applications. This caster features a 4 in. diameter polyurethane wheel, a 1/2-13 threaded stem, and a 275 lb capacity, ensuring durable performance and reliable mobility.
  • Wheel diameter is 4 in.
  • Wheel material is polyurethane and rubber
  • Caster type is stem mount swivel
  • Capacity is 275 lb
  • Bearing type is Delrin
  • Stem type is threaded
  • Stem diameter is 1/2-13
  • Construction is hardened-heat treated
  • Application for industrial and commercial use
  • Finish is zinc chromate