Learn More
Organomation Set of 50 4in. X 19ga. Stainless Steel Needles with a Brass Plated Hub
SDP
Catalog No. NC9361647
Encompass_Preferred
Set of 50 luer lock needles. These needles are 4 inch by 19 gauge, with a 304 stainless steel shaft and a chrome plated brass hub.
Catalog No. NC9361647 Supplier Organomation Supplier No. NA1303
Set of 50 luer lock needles. These needles are 4 inch by 19 gauge, with a 304 stainless steel shaft and a chrome plated brass hub.