Learn More
Gmp Labeling Inc Durable 4x2.5 YLLW Therm Trans Lbl
SDP
Catalog No. NC3483184
Encompass_Preferred
Roll Labels, Thermal Transfer, Chemical Resistant Polyester, 4" x 2.5", Yellow, Permanent Ahdesive, 500 labels/roll
Catalog No. NC3483184 Supplier Gmp Labeling Inc Supplier No. L242TTYLW
Roll Labels, Thermal Transfer, Chemical Resistant Polyester, 4" x 2.5", Yellow, Permanent Ahdesive, 500 labels/roll