Learn More
Abcam Europium Conjugation Kit, 4 x 4UG
Catalog No. NC2195054
Encompass_Preferred
Europium conjugation / labeling in < 4 hrs with 30 secs hands-on time using Europium Conjugation Kit ab269889. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
This item is not returnable. View return policy
Catalog No. NC2195054 Supplier Abcam Supplier No. AB2698894X4UG
Europium conjugation / labeling in < 4 hrs with 30 secs hands-on time using Europium Conjugation Kit ab269889. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement