Learn More
Alkali Scientific Stainless Steel Freezer Racks for 2" Boxes, 4 x 4 Configuration, 16 Box Capacity
SDP
Catalog No. 502336210
Encompass_Preferred
Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-handed doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions (DxHxW): 22 3/16 x 8 7/8x 5 1/2 inches (429 x 225 x 140mm). Compartment Dimensions(DxHxW): 5 3/8 x 2 3/16 x 5 7/16 inches (136 x 55 x 138mm).
Catalog No. 50-233-6210 Supplier Alkali Scientific Supplier No. UFR442
Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left-handed doors. Our freezer racks have been configured to fit all major ultra-low (-80°) freezers on the market; racks are designed for ultra-low upright freezers. Dimensions (DxHxW): 22 3/16 x 8 7/8x 5 1/2 inches (429 x 225 x 140mm). Compartment Dimensions(DxHxW): 5 3/8 x 2 3/16 x 5 7/16 inches (136 x 55 x 138mm).