Learn More
Labrepco Inc Upright Freezer Rack UF-462, Holds 24 Boxes, Stainless Steel Construction, 22.2 in
SDP
Catalog No. NC2878448
Encompass_Preferred
The Upright Freezer Rack by LABREPCO INC is designed for efficient cryogenic storage, featuring stainless steel construction for durability. It holds 24 standard 2-inch cryoboxes in a 4x6 configuration, optimizing space in ultra-low temperature freezers. Integrated handles ensure easy retrieval, making this rack ideal for laboratories looking for organization and accessibility in cold storage solutions.
Catalog No. NC2878448 Supplier Labrepco Inc Supplier No. UF462P
The Upright Freezer Rack by LABREPCO INC is designed for efficient cryogenic storage, featuring stainless steel construction for durability. It holds 24 standard 2-inch cryoboxes in a 4x6 configuration, optimizing space in ultra-low temperature freezers. Integrated handles ensure easy retrieval, making this rack ideal for laboratories looking for organization and accessibility in cold storage solutions.