Learn More
Pickering Laboratories Inc 4X950ML SODIUM ELUENT NA270
SDP
Catalog No. NC9535843
Encompass_Preferred
Sodium Eluant, pH 2.80, case of 4 (950 mL/bottle) Amino Acid Analysis.
Catalog No. NC9535843 Supplier Pickering Laboratories Inc Supplier No. NA270
Sodium Eluant, pH 2.80, case of 4 (950 mL/bottle) Amino Acid Analysis.