Learn More
Teknova 4xYT Broth. 500mL, Sterile. 4 pack.
SDP
Catalog No. 502387275
Encompass
3.2% Tryptone, 2.0% Yeast Extract,1.0% Sodium Chloride
Catalog No. 50-238-7275 Supplier Teknova Supplier No. 2Y10854PK
3.2% Tryptone, 2.0% Yeast Extract,1.0% Sodium Chloride