Learn More
ADAPTIVE SURGICAL LLC Silk Suture, Size: 5/0. Needle: P-3, 13mm. 3/8 Circle Length: 18 in, 12/box
Catalog No. NC9140104
Encompass
640S CP SILK SUTURE, Size: 5/0. Needle: P-3, 13mm. 3/8 Circle Length: 18 in.~45cm. Color: BLACK, 12/box. (Comparable to Ethicon 640G). Supplier will only ship to the following states: IN, KY, VT, ND, OR, WA, NM, TX, MN, AR, MO, MA, WI, CO, NE, HI, OH.
Catalog No. NC9140104 Supplier ADAPTIVE SURGICAL LLC Supplier No. 640S
640S CP SILK SUTURE, Size: 5/0. Needle: P-3, 13mm. 3/8 Circle Length: 18 in.~45cm. Color: BLACK, 12/box. (Comparable to Ethicon 640G). Supplier will only ship to the following states: IN, KY, VT, ND, OR, WA, NM, TX, MN, AR, MO, MA, WI, CO, NE, HI, OH.