Learn More
Roboz Surgical 5-0 SUTURE BLACK SILK
SDP
Catalog No. NC9422522
Encompass
Suture Material,Non-Sterile, 100Yard Spool. Size: 5-0; Silk, Black, Braided, Non-Absorbable
Catalog No. NC9422522 Supplier Roboz Surgical Supplier No. SUT151
Add to Cart
Add to Cart
Suture Material,Non-Sterile, 100Yard Spool. Size: 5-0; Silk, Black, Braided, Non-Absorbable