Learn More
AURICAL LLC Low Level Conductivity Standard 0.5µs/cm 500 mL pouches
Catalog No. NC0074384
Encompass_Preferred
Low Level Conductivity Standard 0.5µs/cm 500 mL pouches for accurate conductivity measurement in purified water. Standard for accurate conductivity measurement in purified water. High accuracy is required for pure water and water for injection in pharmaceutical manufacturing.
Catalog No. NC0074384 Supplier AURICAL LLC Supplier No. 3000.52
Low Level Conductivity Standard 0.5µs/cm 500 mL pouches for accurate conductivity measurement in purified water. Standard for accurate conductivity measurement in purified water. High accuracy is required for pure water and water for injection in pharmaceutical manufacturing.