Learn More
Chemglass Life Sciences Cryogenic Vial, 5.0 mL, Self-Standing, External Thread, Sterile, Racked Nest
SDP
Catalog No. 502733389
Encompass_Preferred
5.0 mL cryogenic vial designed for ultra-low temperature sample storage. The one-piece, self-standing construction preserves barcode and label integrity during cryogenic storage. Sterilized by electron beam and autoclavable, the vial has clear graduations and a white marking area for sample identification.
  • Capacity: 5.0 mL.
  • Manufactured with USP 6 grade raw materials.
  • External thread.
  • Sterilized by e-beam.
  • Packaging: 9 x 9 per rack; 6 racks per case; 486 vials per case.
  • Autoclavable.
  • Recommended storage: gas phase of liquid nitrogen.
  • Non-pyrogenic and free of DNase/RNase.
Catalog No. 50-273-3389 Supplier Chemglass Life Sciences Supplier No. CGN4762006
5.0 mL cryogenic vial designed for ultra-low temperature sample storage. The one-piece, self-standing construction preserves barcode and label integrity during cryogenic storage. Sterilized by electron beam and autoclavable, the vial has clear graduations and a white marking area for sample identification.
  • Capacity: 5.0 mL.
  • Manufactured with USP 6 grade raw materials.
  • External thread.
  • Sterilized by e-beam.
  • Packaging: 9 x 9 per rack; 6 racks per case; 486 vials per case.
  • Autoclavable.
  • Recommended storage: gas phase of liquid nitrogen.
  • Non-pyrogenic and free of DNase/RNase.