Learn More
Chemglass Life Sciences Chemglass 5.0 mL Cryogenic Vial, Self-Standing, Internal Thread, Sterile, Racked (CGN-4766-014)
SDP
Catalog No. 502733339
Encompass_Preferred
This 5.0 mL self-standing cryogenic vial features an internal thread and sterile manufacturing for reliable cryogenic sample storage. Supplied in racked nests for organized handling, the vial is made from USP Class VI grade raw materials and is recommended for storage in the gas phase of liquid nitrogen.
  • Self-standing design for stable upright storage.
  • 5.0 mL nominal volume.
  • Internal thread to secure compatible caps.
  • Sterile packaging to reduce contamination risk.
  • Supplied in 9 x 9 racks; six racks per case (486 vials per case).
  • Manufactured from USP Class VI grade materials suitable for cryogenic use.
  • Recommended for storage in the gas phase of liquid nitrogen.
Catalog No. 50-273-3339 Supplier Chemglass Life Sciences Supplier No. CGN4766014
This 5.0 mL self-standing cryogenic vial features an internal thread and sterile manufacturing for reliable cryogenic sample storage. Supplied in racked nests for organized handling, the vial is made from USP Class VI grade raw materials and is recommended for storage in the gas phase of liquid nitrogen.
  • Self-standing design for stable upright storage.
  • 5.0 mL nominal volume.
  • Internal thread to secure compatible caps.
  • Sterile packaging to reduce contamination risk.
  • Supplied in 9 x 9 racks; six racks per case (486 vials per case).
  • Manufactured from USP Class VI grade materials suitable for cryogenic use.
  • Recommended for storage in the gas phase of liquid nitrogen.