Learn More
Trajan Scientific L-MARK Brand 5.0mL Syringe, FN, GT, 26g, NDL Length 56mm, Pt. Style 5, Plunger Tip PTFE, Single
Catalog No. 50635281
Encompass_Preferred
L-MARK Brand Syringe, 5.0-mL, FN, GT, PTFE, 26g, 56mm, Pt.5, Headspace; Replacement Plunger: LMK.2636043 Made in Germany.
Catalog No. 50-635-281 Supplier Trajan Scientific Supplier No. LMK.2620051
L-MARK Brand Syringe, 5.0-mL, FN, GT, PTFE, 26g, 56mm, Pt.5, Headspace; Replacement Plunger: LMK.2636043 Made in Germany.