Learn More
Sklar Instruments Single Use, Econo-Sterile, Crile Forceps, Sklar 5-1/2 in., Blunt, Straight, Serrated, Stainless Steel, Disposable. CS of 25.
SDP
Catalog No. 501094266 Shop All Sklar Instruments Products
Encompass_Preferred
Single Use, Econo-Sterile, Crile Forceps, Sklar, 5-1/2 in., Blunt, Straight, Serrated, Stainless Steel, Disposable. CS of 25. Crile Hemostatic Forceps, also known as a Crile Hemostat, are most commonly used for clamping blood vessels or tissue before cauterization or ligation.
Catalog No. 50-109-4266 Supplier Sklar Instruments Supplier No. 962551
Single Use, Econo-Sterile, Crile Forceps, Sklar, 5-1/2 in., Blunt, Straight, Serrated, Stainless Steel, Disposable. CS of 25. Crile Hemostatic Forceps, also known as a Crile Hemostat, are most commonly used for clamping blood vessels or tissue before cauterization or ligation.