Learn More
Sklar Instruments Single Use, Econo-Sterile, Metzenbaum-Lahey Dissecting Scissors,Sklar -1/2 in., Blunt/Blunt, Curved, Smooth, Stainless Steel, Disposable, CS of 25.
SDP
Catalog No. 501094361 Shop All Sklar Instruments Products
Encompass_Preferred
Single Use, Econo-Sterile, Metzenbaum-Lahey Dissecting Scissors,Sklar, 5-1/2 in., Blunt/Blunt, Curved, Smooth, Stainless Steel, Disposable, CS of 25. Metzenbaum Dissecting Scissors are most commonly used for dissecting and cutting tissue.
Catalog No. 50-109-4361 Supplier Sklar Instruments Supplier No. 962720
Single Use, Econo-Sterile, Metzenbaum-Lahey Dissecting Scissors,Sklar, 5-1/2 in., Blunt/Blunt, Curved, Smooth, Stainless Steel, Disposable, CS of 25. Metzenbaum Dissecting Scissors are most commonly used for dissecting and cutting tissue.