Learn More
Sklar Instruments Single Use, Econo-Sterile, Metzenbaum-Lahey Dissecting Scissors,Sklar -1/2 in., Blunt/Blunt, Curved, Smooth, Stainless Steel, Disposable, CS of 50.
SDP
Catalog No. 501094362 Shop All Sklar Instruments Products
Encompass_Preferred
Single Use, Econo-Sterile, Metzenbaum-Lahey Dissecting Scissors,Sklar, 5-1/2 in., Blunt/Blunt, Curved, Smooth, Stainless Steel, Disposable, CS of 50. Metzenbaum Dissecting Scissors are most commonly used for dissecting and cutting tissue.
Catalog No. 50-109-4362 Supplier Sklar Instruments Supplier No. 962721
Single Use, Econo-Sterile, Metzenbaum-Lahey Dissecting Scissors,Sklar, 5-1/2 in., Blunt/Blunt, Curved, Smooth, Stainless Steel, Disposable, CS of 50. Metzenbaum Dissecting Scissors are most commonly used for dissecting and cutting tissue.