Learn More
US Plastic Corporation 5-1/2 Gallon Nalgene Wide Mouth LDPE Carboy Modified by Tamco with a Fast Draw Off Spigot
SDP
Supplier:  US Plastic Corporation 65033
Encompass
Our finest polyethylene carboy, featuring a spigot that has 3/4"-16 UNF with a 1" long thread, flow rate of 1.18 gpm. Is lab quality, heavy duty, leakproof, and resistant to most chemicals.
Catalog No. NC1837028