Learn More
Sklar Instruments Forceps, Kelly Hemostatic Forceps, Sklar 5-1/2 in., Curved, Serrated, Delicate, Finger Rings, Stainless Steel, EA.
SDP
Catalog No. 501097971 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Kelly Hemostatic Forceps, Sklar, 5-1/2 in., Curved, Serrated, Delicate, Finger Rings, Stainless Steel, EA. Sklar Kelly Hemostatic Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. They may also be used for soft tissue dissection. They are available curved or straight with a ratcheted finger ring handle. The jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws. This product is curved with serrated tips and a length of 5-1/2 inches. They also feature a delicate pattern.
Catalog No. 50-109-7971 Supplier Sklar Instruments Supplier No. 181652
Forceps, Kelly Hemostatic Forceps, Sklar, 5-1/2 in., Curved, Serrated, Delicate, Finger Rings, Stainless Steel, EA. Sklar Kelly Hemostatic Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue. They may also be used for soft tissue dissection. They are available curved or straight with a ratcheted finger ring handle. The jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws. This product is curved with serrated tips and a length of 5-1/2 inches. They also feature a delicate pattern.