Learn More
Sklar Instruments Scissors, Sklar Mayo Dissecting Scissors, Sklar 5-1/2 in., Blunt/Blunt, Straight, Razor, Stainless Steel, Reusable. EA.
SDP
Catalog No. 501093407 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Sklarhone Mayo Dissecting Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Straight, Razor, Stainless Steel, Reusable. EA. Sklarhone Mayo Dissecting Scissors are a variation of standard Mayo Scissors that feature one knife-like razor sharp edge and one standard scissors edge. This allows the user to make the sharp incisions of a knife with the cutting ability of scissors, while catching skin and pulls are minimized. These scissors are a versatile instrument that can be used in multiple procedures and settings. They are available straight or curved, in a variety of lengths, and have a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference. Identified by a dual black finger ring handle, this product is straight, and an overall length of 5-1/2 inches.
Catalog No. 50-109-3407 Supplier Sklar Instruments Supplier No. 153325
Scissors, Sklarhone Mayo Dissecting Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Straight, Razor, Stainless Steel, Reusable. EA. Sklarhone Mayo Dissecting Scissors are a variation of standard Mayo Scissors that feature one knife-like razor sharp edge and one standard scissors edge. This allows the user to make the sharp incisions of a knife with the cutting ability of scissors, while catching skin and pulls are minimized. These scissors are a versatile instrument that can be used in multiple procedures and settings. They are available straight or curved, in a variety of lengths, and have a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference. Identified by a dual black finger ring handle, this product is straight, and an overall length of 5-1/2 inches.