Learn More
Sklar Instruments Scissors, Sklar Operating Scissors, Sklar 5-1/2 in., Sharp/Sharp, 1 Knifelike Razor Edge and 1 Micro-serrated Edge, Stainless Steel, EA.
SDP
Catalog No. 501093457 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Sklarcut Operating Scissors, Sklar, 5-1/2 in., Sharp/Sharp, 1 Knifelike Razor Edge and 1 Micro-serrated Edge, Stainless Steel, EA. Sklarcut Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Sklarcut scissors feature micro-serrations on one blade to reduce trauma when cutting both delicate and course tissue. These serrations assist in securely holding the tissue during cutting and minimizes tissue from slipping so the most accurate and clean cuts can be made.
Catalog No. 50-109-3457 Supplier Sklar Instruments Supplier No. 153500
Scissors, Sklarcut Operating Scissors, Sklar, 5-1/2 in., Sharp/Sharp, 1 Knifelike Razor Edge and 1 Micro-serrated Edge, Stainless Steel, EA. Sklarcut Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument. With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze. Sklarcut scissors feature micro-serrations on one blade to reduce trauma when cutting both delicate and course tissue. These serrations assist in securely holding the tissue during cutting and minimizes tissue from slipping so the most accurate and clean cuts can be made.