Learn More
Sklar Instruments Scissors, Sklar Extra Delicate Mayo Dissecting Scissors, Sklar 5-1/2 in., Blunt/Blunt, Straight, Smooth, Extra Light and Extra Delicate (Sklar XD), Stainless Steel, EA.
SDP
Catalog No. 501093694 Shop All Sklar Instruments Products
Encompass_Preferred
Scissors, Sklarlite Extra Delicate Mayo Dissecting Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Straight, Smooth, Extra Light and Extra Delicate (SklarLite XD), Stainless Steel, EA. Sklarlite XD Mayo Dissecting Scissors are an extra light and extra delicate version of the standard Mayo scissors that offer the surgeon the highest degree of precision with the least amount of fatigue, while maintaining structural integrity and strength. Mayo Dissecting Scissors are a versatile instrument that can be used in multiple procedures and settings. They are available straight or curved in a variety of lengths with a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference.
Catalog No. 50-109-3694 Supplier Sklar Instruments Supplier No. 231168
Scissors, Sklarlite Extra Delicate Mayo Dissecting Scissors, Sklar, 5-1/2 in., Blunt/Blunt, Straight, Smooth, Extra Light and Extra Delicate (SklarLite XD), Stainless Steel, EA. Sklarlite XD Mayo Dissecting Scissors are an extra light and extra delicate version of the standard Mayo scissors that offer the surgeon the highest degree of precision with the least amount of fatigue, while maintaining structural integrity and strength. Mayo Dissecting Scissors are a versatile instrument that can be used in multiple procedures and settings. They are available straight or curved in a variety of lengths with a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference.