Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-(1,3-Benzodioxol-5-yl)-2,4-pentadienoic acid

Catalog No. 7580781G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
75-807-81G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 75-807-81G Supplier Sigma Aldrich Supplier No. 7580781G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-propylphenol can be used as:
  • A ligand to prepare iridium complex for the study of equilibria between iridium hydride alkylidene structures and their corresponding hydride olefin isomers
  • A model compound in the study of pt/c-catalyzed h-d exchange reactions of aromatic compounds using d2o

Specifications

CAS 5285-18-7
Melting Point 218°C to 223°C
Molecular Formula C12H10O4
Linear Formula C12H10O4
MDL Number MFCD00047282
Quantity 25 g
Synonym 5-(3,4-Methylenedioxyphenyl)-2,4-pentadienoic acid; Piperic acid
Molecular Weight (g/mol) 218.21
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.