Learn More
Medchemexpress LLC 5-(1,3-benzodioxol-5-ylmethylene)-2,4-thiazolidinedione | 6318-41-8 | MFCD18382117 | 99.0% | 249.24 g/mol | C11H7NO4S | 1 ML
SDP
Catalog No. 5000227716
Encompass_Preferred
AS-041164 is a potent, selective, orally active PI3Kγ inhibitor supplied either as a ready-to-use solution (10 mM in DMSO, 1 mL) or as a solid in multiple mg sizes for research applications investigating PI3Kγ signaling and inflammatory responses.
  • Potent, selective PI3Kγ inhibition with IC50 ≈ 70 nM.
  • Available as 10 mM solution (1 mL) or solids in multiple milligram sizes.
  • High purity (≈99.0%) suitable for research use.
  • Soluble in DMSO, convenient for in vitro assays.
  • Store protected from light; in solvent conditions extend stability at low temperatures.
Catalog No. 50-002-27716 Supplier Medchemexpress LLC Supplier No. HY11852110MM 1ML
AS-041164 is a potent, selective, orally active PI3Kγ inhibitor supplied either as a ready-to-use solution (10 mM in DMSO, 1 mL) or as a solid in multiple mg sizes for research applications investigating PI3Kγ signaling and inflammatory responses.
  • Potent, selective PI3Kγ inhibition with IC50 ≈ 70 nM.
  • Available as 10 mM solution (1 mL) or solids in multiple milligram sizes.
  • High purity (≈99.0%) suitable for research use.
  • Soluble in DMSO, convenient for in vitro assays.
  • Store protected from light; in solvent conditions extend stability at low temperatures.