Learn More
Sklar Instruments Forceps, Econo Backhaus Towel Forceps, Sklar 5-1/4 in., 1x1 Prongs, Curved, Pointed, Finger Rings, Disposable, Stainless Steel, BX/12.
SDP
Catalog No. 501098093 Shop All Sklar Instruments Products
Encompass_Preferred
Forceps, Econo Backhaus Towel Forceps, Sklar, 5-1/4 in., 1x1 Prongs, Curved, Pointed, Finger Rings, Disposable, Stainless Steel, BX/12. Sklar Backhaus Towel Forceps are a perforating clamp used for securing towels or drapes, grasping tissue, and holding or reducing small bone fractures. Econo instruments are Sklar's floor-grade, disposable instruments. Made in Pakistan, they offer the greatest utility at the lowest cost. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is curved with perforating 1x1 prongs and an overall length of 5-1/4 inches. They are sold in boxes of 12.
Catalog No. 50-109-8093 Supplier Sklar Instruments Supplier No. 21608
Forceps, Econo Backhaus Towel Forceps, Sklar, 5-1/4 in., 1x1 Prongs, Curved, Pointed, Finger Rings, Disposable, Stainless Steel, BX/12. Sklar Backhaus Towel Forceps are a perforating clamp used for securing towels or drapes, grasping tissue, and holding or reducing small bone fractures. Econo instruments are Sklar's floor-grade, disposable instruments. Made in Pakistan, they offer the greatest utility at the lowest cost. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is curved with perforating 1x1 prongs and an overall length of 5-1/4 inches. They are sold in boxes of 12.