Learn More
Medchemexpress LLC 5-(1-methyl-1H-pyrazol-4-yl)-N-(2-methyl-5-(3-(trifluoromethyl)benzamido)phenyl)nicotinamide | 2095849-04-8 | MFCD31813597 | 98.0% | 479.45 g/mol | C25H20F3N5O2 | 50 MG
SDP
Catalog No. 5000245401
Encompass_Preferred
CSF1R-IN-1 is a potent small-molecule inhibitor of the colony stimulating factor 1 receptor (CSF1R) with a reported IC50 of 0.5 nM. It shows good intestinal permeability in Caco-2 assays and favorable oral pharmacokinetics in mice. The compound is supplied as a solid for research use and includes standard storage recommendations and available solution formulations.
  • Potent CSF1R inhibition (IC50 0.5 nM).
  • Molecular formula C25H20F3N5O2; molecular weight 479.45 g/mol.
  • High purity (98.04%).
  • White to off-white solid physical form.
  • Stable storage: powder -20°C for up to 3 years; in solvent -80°C for 6 months, -20°C for 1 month.
  • Available in multiple sizes and as a 10 mM solution in DMSO.
Catalog No. 50-002-45401 Supplier Medchemexpress LLC Supplier No. HY10177450MG
CSF1R-IN-1 is a potent small-molecule inhibitor of the colony stimulating factor 1 receptor (CSF1R) with a reported IC50 of 0.5 nM. It shows good intestinal permeability in Caco-2 assays and favorable oral pharmacokinetics in mice. The compound is supplied as a solid for research use and includes standard storage recommendations and available solution formulations.
  • Potent CSF1R inhibition (IC50 0.5 nM).
  • Molecular formula C25H20F3N5O2; molecular weight 479.45 g/mol.
  • High purity (98.04%).
  • White to off-white solid physical form.
  • Stable storage: powder -20°C for up to 3 years; in solvent -80°C for 6 months, -20°C for 1 month.
  • Available in multiple sizes and as a 10 mM solution in DMSO.