Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carbaldehyde, 97%, Thermo Scientific™

Catalog No. KM05281CB
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KM05281CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KM05281CB Supplier Thermo Scientific Chemicals Supplier No. KM05281CB
Add to Cart

Chemical Identifiers

CAS 175202-93-4
Molecular Formula C10H7F3N2OS
Molecular Weight (g/mol) 260.234
MDL Number MFCD00052163
InChI Key ITUYPRXQRPMRTL-UHFFFAOYSA-N
Synonym 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl thiophene-2-carbaldehyde, 5-1-methyl-5-trifluoromethyl pyrazol-3-yl thiophene-2-carbaldehyde, 2-1-methyl-5-trifluoromethyl pyrazol-3-yl-thiophene-5-carboxaldehyde, 2-1-methyl-5-trifluoromethyl pyrazol-3-yl thiophene-5-carboxaldehyde, 2-1-methyl-5-trifluoromethyl pyrazol-3-yl thiophene-5-carbaldehyde, 2-thiophenecarboxaldehyde,5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl, 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl-2-thiophenecarbaldehyde, 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl-2-thiophenecarboxaldehyde, 5-1-methyl-5-trifluoromethyl-1h-pyrazole-3-yl thiophene-2-carbaldehyde
PubChem CID 2775667
IUPAC Name 5-[1-methyl-5-(trifluoromethyl)pyrazol-3-yl]thiophene-2-carbaldehyde
SMILES CN1C(=CC(=N1)C2=CC=C(S2)C=O)C(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 260.24
Percent Purity 97%
Chemical Name or Material 5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.