Learn More
ANALYTICAL SCIENTIFIC INSTRUMENTS US INC 5:1 Split Ratio, Analytical Binary Fixed Flow Splitter, 0.1 to 5 mL/min Inlet Flow
SDP
Catalog No. NC2604723
Encompass
The ASI 5:1 Split Ratio Analytical Binary Fixed Flow Splitter is engineered for precise post-column applications in chromatography. Its advanced fluid resistor technology guarantees stable split ratios, increasing reliability and efficiency in analytical processes. With a flow range of 0.1 to 5 mL/min and high-pressure tolerance, it suits demanding laboratory environments. Its compatibility with standard HPLC fittings and robust materials differentiate it, ensuring optimal performance in various chemical conditions.
Catalog No. NC2604723 Supplier ANALYTICAL SCIENTIFIC INSTRUMENTS US INC Supplier No. 620PO1011
The ASI 5:1 Split Ratio Analytical Binary Fixed Flow Splitter is engineered for precise post-column applications in chromatography. Its advanced fluid resistor technology guarantees stable split ratios, increasing reliability and efficiency in analytical processes. With a flow range of 0.1 to 5 mL/min and high-pressure tolerance, it suits demanding laboratory environments. Its compatibility with standard HPLC fittings and robust materials differentiate it, ensuring optimal performance in various chemical conditions.