Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,10,15,20-Tetrakis[3,5-bis(per-O-methyl-alpha-cyclodextrin-6-yloxy)phenyl]-21H,23H-porphine, TCI America™
SDP

Catalog No. T245210MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T245210MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T245210MG Supplier TCI America Supplier No. T245210MG
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

Molecular Formula C468H766N4O240
Synonym 5,10,15,20-Tetrakis[3,5-bis(per-O-methyl-alpha-cyclodextrin-6-yloxy)phenyl]porphyrin

Specifications

Color Purple
Quantity 10 mg
Formula Weight 10289.06
Physical Form Crystalline Powder
Chemical Name or Material 5,10,15,20-Tetrakis[3,5-bis(per-O-methyl-alpha-cyclodextrin-6-yloxy)phenyl]-21H,23H-porphine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.