Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,10,15,20-Tetrakis(3,5-dimethoxyphenyl)porphyrin 95.0+%, TCI America™
SDP

Catalog No. T1832100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1832100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1832100MG Supplier TCI America Supplier No. T1832100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 74684-34-7
Molecular Formula C52H46N4O8
Molecular Weight (g/mol) 854.96
MDL Number MFCD02093501
InChI Key MOOXLDFVIJGBRF-UHFFFAOYSA-N
Synonym 5,10,15,20-Tetrakis(3,5-dimethoxyphenyl)-21H,23H-porphine
PubChem CID 471972
IUPAC Name 5,10,15,20-tetrakis(3,5-dimethoxyphenyl)-21,22-dihydroporphyrin
SMILES COC1=CC(=CC(=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC(=CC(=C7)OC)OC)C8=CC(=CC(=C8)OC)OC)C9=CC(=CC(=C9)OC)OC)N3)OC

Specifications

Color Blue-Purple
Quantity 100 mg
Formula Weight 854.96
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5,10,15,20-Tetrakis(3,5-dimethoxyphenyl)porphyrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.