Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,10,15,20-Tetrakis(4-aminophenyl)porphyrin 95.0+%, TCI America™
SDP

Catalog No. T1494100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1494100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1494100MG Supplier TCI America Supplier No. T1494100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22112-84-1
Molecular Formula C44H34N8
Molecular Weight (g/mol) 674.81
MDL Number MFCD00191497
InChI Key WLCKPHYUFDYULK-UHFFFAOYSA-N
Synonym 5,10,15,20-Tetrakis(4-aminophenyl)-21H,23H-porphine
PubChem CID 168023
IUPAC Name 4-[7,12,17-tris(4-aminophenyl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11(23),12,14,16(22),17,19-undecaen-2-yl]aniline
SMILES NC1=CC=C(C=C1)C1=C2NC(C=C2)=C(C2=NC(C=C2)=C(C2=NC(C=C2)=C(C2=CC=C1N2)C1=CC=C(N)C=C1)C1=CC=C(N)C=C1)C1=CC=C(N)C=C1

Specifications

Color Blue-Purple
Quantity 100 mg
Formula Weight 674.81
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5,10,15,20-Tetrakis(4-aminophenyl)porphyrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.