Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,10,15,20-tetrakis(4-hydroxyphenyl)porphyrin | 51094-17-8 | 98.0% | 678.73 g·mol⁻1 | C44H30N4O4 | 250 MG
SDP

Catalog No. 5000303547
Encompass_Preferred

5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin is a porphyrin-based organic compound used as a biochemical reagent in life-science research. It is commonly employed in photophysical studies, sensor and catalysis research, and other laboratory applications that require a well-characterized porphyrin scaffold.

  • High purity (98.0%) suitable for research applications.
  • Solid, brown-to-black appearance for easy handling and storage.
  • Chemical formula C44H30N4O4 and molecular weight 678.73 g·mol⁻1.
  • Available in multiple pack sizes for small-scale experiments.
  • Well-suited for photophysical and porphyrin chemistry studies.

Catalog No. 50-003-03547 Supplier Medchemexpress LLC Supplier No. 250MGHYW035140
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin is a porphyrin-based organic compound used as a biochemical reagent in life-science research. It is commonly employed in photophysical studies, sensor and catalysis research, and other laboratory applications that require a well-characterized porphyrin scaffold.

  • High purity (98.0%) suitable for research applications.
  • Solid, brown-to-black appearance for easy handling and storage.
  • Chemical formula C44H30N4O4 and molecular weight 678.73 g·mol⁻1.
  • Available in multiple pack sizes for small-scale experiments.
  • Well-suited for photophysical and porphyrin chemistry studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.