Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,10,15,20-tetrakis(4-hydroxyphenyl)porphyrin | 51094-17-8 | MFCD00191501 | 98.0% | 678.73 g/mol | C44H30N4O4 | 100 MG
SDP

Catalog No. 5000299310
Encompass_Preferred

5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin is a porphyrin-class organic reagent used in life-science and materials research. It contains four hydroxyphenyl groups at the meso positions of the porphyrin ring and is supplied as a solid suitable for photophysical studies, synthetic chemistry, and advanced materials applications.

  • High purity (98.0%).
  • Molecular weight 678.73 g/mol.
  • Chemical formula C44H30N4O4.
  • Solid, brown to black appearance.
  • Available in multiple package sizes for flexible use.
  • Protect from light; in solvent store -80 °C for long term or -20 °C for shorter term.
  • Suitable for photophysical studies, porphyrin synthesis, and materials research.

Catalog No. 50-002-99310 Supplier Medchemexpress LLC Supplier No. HYW035140100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin is a porphyrin-class organic reagent used in life-science and materials research. It contains four hydroxyphenyl groups at the meso positions of the porphyrin ring and is supplied as a solid suitable for photophysical studies, synthetic chemistry, and advanced materials applications.

  • High purity (98.0%).
  • Molecular weight 678.73 g/mol.
  • Chemical formula C44H30N4O4.
  • Solid, brown to black appearance.
  • Available in multiple package sizes for flexible use.
  • Protect from light; in solvent store -80 °C for long term or -20 °C for shorter term.
  • Suitable for photophysical studies, porphyrin synthesis, and materials research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.