Learn More
Medchemexpress LLC 5,10,15,20-tetrakis(4-hydroxyphenyl)porphyrin | 51094-17-8 | MFCD00191501 | 98.0% | 678.73 g/mol | C44H30N4O4 | 5 G
SDP
Catalog No. 5000299460
Encompass_Preferred
5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin is an organic porphyrin compound supplied for research use. It serves as a multifunctional reagent for studies in photochemistry, porphyrin chemistry, sensor development, and materials functionalization, offering predictable porphyrin core reactivity and well-defined spectroscopic properties.
  • High purity (98.0%).
  • Molecular weight 678.73 g/mol; formula C44H30N4O4.
  • Solid, brown to black appearance; store protected from light.
  • Suitable for photochemical and spectroscopic studies.
  • Useful as a building block for porphyrin-based materials and sensors.
  • Available in laboratory pack sizes from 100 mg to 5 g (larger sizes by quote).
Catalog No. 50-002-99460 Supplier Medchemexpress LLC Supplier No. HYW0351405G
5,10,15,20-Tetrakis(4-hydroxyphenyl)porphyrin is an organic porphyrin compound supplied for research use. It serves as a multifunctional reagent for studies in photochemistry, porphyrin chemistry, sensor development, and materials functionalization, offering predictable porphyrin core reactivity and well-defined spectroscopic properties.
  • High purity (98.0%).
  • Molecular weight 678.73 g/mol; formula C44H30N4O4.
  • Solid, brown to black appearance; store protected from light.
  • Suitable for photochemical and spectroscopic studies.
  • Useful as a building block for porphyrin-based materials and sensors.
  • Available in laboratory pack sizes from 100 mg to 5 g (larger sizes by quote).