Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine iron(III) chloride

Catalog No. 303038100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-303-8100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 30-303-8100MG Supplier Sigma Aldrich Supplier No. 303038100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine iron(III) chloride (FeTMPPCl) is a metalloporphyrin.FeTMPPCI may be used to form interfacial molecular assemblies of metalloporphyrins. It has been used in the delignification of wood chips and pulps.

Specifications

CAS 36995-20-7
Molecular Formula C48H36ClFeN4O4
Linear Formula C48H36ClFeN4O4
MDL Number MFCD00012405
Quantity 100 mg
Molecular Weight (g/mol) 824.12
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.