Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin 97.0+%, TCI America™
SDP

Catalog No. T13601G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13601G 1 g
T1360100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13601G Supplier TCI America Supplier No. T13601G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22112-78-3
Molecular Formula C48H38N4O4
Molecular Weight (g/mol) 734.856
MDL Number MFCD00012072
InChI Key SKXWDGDNWBYACJ-UHFFFAOYSA-N
Synonym 5,10,15,20-tetrakis 4-methoxyphenyl-21h,23h-porphine, 5,10,15,20-tetrakis 4-methoxyphenyl porphyrin, tetra p-methoxyphenyl porphyrin, 21h,23h-porphine, 5,10,15,20-tetrakis 4-methoxyphenyl, 5,10,15,20-tetrakis 4-methoxyphenyl-21,22-dihydroporphyrin, 2,7,12,17-tetrakis 4-methoxyphenyl-21,22,23,24-tetraazapentacyclo 16.2.1.1 3 ,?.1?, 1 1 .1 1 3 , 1 ? tetracosa-1,3,5,7,9,11 23 ,12,14,16,18 21 ,19-undecaene, 4-methoxy-1-7,12,17-tris 4-methoxyphenyl-21,22,23,24-tetraazapentacyclo 16.2 .1.1<3,6>.1<8,11>.1<13,16> tetracosa-1,3,5,7,9,11 23 ,12,14,16,18 21 ,19-undec aen-2-yl benzene, acmc-209fs6, tetrakis p-methoxyphenyl porphyrin, meso-tetra 4-methoxyphenyl porphine
PubChem CID 140886
IUPAC Name 5,10,15,20-tetrakis(4-methoxyphenyl)-21,22-dihydroporphyrin
SMILES COC1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=C(C=C7)OC)C8=CC=C(C=C8)OC)C9=CC=C(C=C9)OC)N3

Specifications

Color Blue-Purple
Quantity 1 g
Formula Weight 734.86
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.