Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrinato]cobalt(II) 96.0+%, TCI America™
SDP

Catalog No. T18615G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18615G 5 g
T18611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18615G Supplier TCI America Supplier No. T18615G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 28903-71-1
Molecular Formula C48H38CoN4O4
Molecular Weight (g/mol) 793.789
MDL Number MFCD00010724
InChI Key RGOMRGJUWGSAHF-UHFFFAOYSA-N
Synonym tetra 4-methoxyphenyl porphyrine cobalt complex
PubChem CID 67369205
IUPAC Name cobalt;5,10,15,20-tetrakis(4-methoxyphenyl)-21,22-dihydroporphyrin
SMILES COC1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=C(C=C7)OC)C8=CC=C(C=C8)OC)C9=CC=C(C=C9)OC)N3.[Co]

Specifications

Color Blue-Purple
Quantity 5 g
Formula Weight 791.77
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrinato]cobalt(II)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.