Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,10,15,20-tetrakis(p-tolyl)porphyrin | 14527-51-6 | MFCD00239499 | 97.0% | 670.84 | C48H38N4 | 1 G
SDP

Catalog No. 5000299352
Encompass_Preferred

5,10,15,20-Tetrakis(p-tolyl)porphyrin is an organic porphyrin reagent supplied as a solid for research applications. It functions as a building block in organic synthesis and is used in photophysical studies, supramolecular chemistry, and catalyst development where substituted porphyrin scaffolds are required.

  • Porphyrin scaffold for photophysical and catalytic studies.
  • Molecular formula C48H38N4 and molecular weight 670.84 g·mol⁻1.
  • High purity (97.0%) suitable for research applications.
  • Solid powder form, stable when protected from light and oxygen.
  • Storage: 4°C, protect from light, store under inert gas; in solution store at -20°C to -80°C for long-term use.
  • Available in small research pack sizes for laboratory workflows.

Catalog No. 50-002-99352 Supplier Medchemexpress LLC Supplier No. HYW0351331G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5,10,15,20-Tetrakis(p-tolyl)porphyrin is an organic porphyrin reagent supplied as a solid for research applications. It functions as a building block in organic synthesis and is used in photophysical studies, supramolecular chemistry, and catalyst development where substituted porphyrin scaffolds are required.

  • Porphyrin scaffold for photophysical and catalytic studies.
  • Molecular formula C48H38N4 and molecular weight 670.84 g·mol⁻1.
  • High purity (97.0%) suitable for research applications.
  • Solid powder form, stable when protected from light and oxygen.
  • Storage: 4°C, protect from light, store under inert gas; in solution store at -20°C to -80°C for long-term use.
  • Available in small research pack sizes for laboratory workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.