Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,10,15,20-Tetrakis(p-tolyl)porphyrin 98.0+%, TCI America™
SDP

Catalog No. T30635G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T30635G Supplier TCI America Supplier No. T30635G

missing translation for 'validMainframeMsg'

Specifications

CAS 14527-51-6
Melting Point 300°C
Color Purple
Molecular Formula C48H38N4
MDL Number MFCD00239499
Quantity 5 g
Synonym 5,10,15,20-Tetrakis(4-methylphenyl)porphyrin, 5,10,15,20-Tetrakis(p-tolyl)-21H,23H-porphine, 5,10,15,20-Tetrakis(4-methylphenyl)-21H,23H-porphine
InChI Key XNOMLEHGHNPLMW-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)C2=C3C=CC(=C(C4=CC=C(N4)C(=C5C=CC(=N5)C(=C6C=CC2=N6)C7=CC=C(C=C7)C)C8=CC=C(C=C8)C)C9=CC=C(C=C9)C)N3
IUPAC Name 5,10,15,20-tetrakis(4-methylphenyl)-21,22-dihydroporphyrin
Molecular Weight (g/mol) 670.86
PubChem CID 139774
Formula Weight 670.86
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5,10,15,20-Tetrakis(p-tolyl)porphyrin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.