Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin

Catalog No. 252921100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-292-1100MG 100 mg
25-292-11G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-292-1100MG Supplier Sigma Aldrich Supplier No. 252921100MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

5,10,15,20-Tetrakis(pentafluorophenyl)porphyrin has been used as a biomarker in a study. 1

Specifications

CAS 25440-14-6
Molecular Formula C44H10F20N4
Linear Formula C44H10F20N4
Quantity 100 mg
MDL Number MFCD00010032
Molecular Weight (g/mol) 974.55
Percent Purity ≥90.0% (HPLC)
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.