Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,10,15,20-tetraphenyl-21H,23H-porphine cobalt(II) | 14172-90-8 | MFCD00010725 | ≥98.0% | 671.65 g/mol | C44H28CoN4 | 5 G
SDP

Catalog No. 5000210639
Encompass_Preferred

5,10,15,20-tetraphenyl-21H,23H-porphine cobalt(II) is a cobalt porphyrin reagent used as an organometallic catalyst and biochemical reagent in organic synthesis and research applications. It is supplied as a high-purity solid and is typically stored protected from light and under inert atmosphere.

  • high purity suitable for research use.
  • available in multiple pack sizes, including 5 g.
  • CAS number 14172-90-8 for unambiguous identification.
  • storage: keep at 4°C, protect from light, store under nitrogen; in solvent, store at -80°C up to 6 months.
  • commonly used as a catalyst and in porphyrin chemistry applications.

Catalog No. 50-002-10639 Supplier Medchemexpress LLC Supplier No. HYW0730135G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5,10,15,20-tetraphenyl-21H,23H-porphine cobalt(II) is a cobalt porphyrin reagent used as an organometallic catalyst and biochemical reagent in organic synthesis and research applications. It is supplied as a high-purity solid and is typically stored protected from light and under inert atmosphere.

  • high purity suitable for research use.
  • available in multiple pack sizes, including 5 g.
  • CAS number 14172-90-8 for unambiguous identification.
  • storage: keep at 4°C, protect from light, store under nitrogen; in solvent, store at -80°C up to 6 months.
  • commonly used as a catalyst and in porphyrin chemistry applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.