Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,10-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphtho[1,2-c:5,6-c']bis([1,2,5]thiadiazole) 98.0+%, TCI America™
SDP

Catalog No. B5470100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5470100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B5470100MG Supplier TCI America Supplier No. B5470100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1467776-41-5
Molecular Formula C22H26B2N4O4S2
Molecular Weight (g/mol) 496.214
MDL Number MFCD28963789
InChI Key VZCYKOASVMGCSP-UHFFFAOYSA-N
Synonym Naphtho[1,2-c:5,6-c′]bis([1,2,5]thiadiazole)-5,10-diboronic Acid Bis(pinacol) Ester
PubChem CID 100912254
IUPAC Name 5,10-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-[2,1,3]benzothiadiazolo[5,4-e][2,1,3]benzothiadiazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C(C4=NSN=C34)B5OC(C(O5)(C)C)(C)C)C6=NSN=C26

Specifications

Melting Point 382°C
Quantity 100 mg
Formula Weight 496.21
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5,10-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphtho[1,2-c:5,6-c′]bis([1,2,5]thiadiazo
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.