Learn More
Roboz Surgical #5 15 DEG DUMONT INOX 10X.06MM
SDP
Catalog No. NC9265019
Encompass
Dumont Tweezers; Pattern #5; 15 Degrees Up; .10 X .06mm Tip Size; 105mm Length; Material: Inox
Catalog No. NC9265019 Supplier Roboz Surgical Supplier No. RS5045A
Dumont Tweezers; Pattern #5; 15 Degrees Up; .10 X .06mm Tip Size; 105mm Length; Material: Inox