Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,15-diphenyl-10,20-di(pyridin-4-yl)porphyrin | 71410-72-5 | MFCD16628185 | ≥98.0% | 616.71 g/mol | C42H28N6 | 250 MG
SDP

Catalog No. 5000209292
Encompass_Preferred

5,15-diphenyl-10,20-di(pyridin-4-yl)porphyrin is a substituted porphyrin used as an organic compound and biochemical reagent for life-science research. It is suitable for material studies and synthetic applications that require high-purity porphyrins.

  • Purity ≥98.0%
  • CAS number 71410-72-5
  • Chemical formula C42H28N6
  • Molecular weight 616.71 g/mol
  • Available sizes 100 mg, 250 mg, 500 mg, 1 g, 5 g, 10 g

Catalog No. 50-002-09292 Supplier Medchemexpress LLC Supplier No. 250MGHYW075433
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5,15-diphenyl-10,20-di(pyridin-4-yl)porphyrin is a substituted porphyrin used as an organic compound and biochemical reagent for life-science research. It is suitable for material studies and synthetic applications that require high-purity porphyrins.

  • Purity ≥98.0%
  • CAS number 71410-72-5
  • Chemical formula C42H28N6
  • Molecular weight 616.71 g/mol
  • Available sizes 100 mg, 250 mg, 500 mg, 1 g, 5 g, 10 g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.