Learn More
Ace Glass, Inc. 5/16" ID tubing hose connection on 3mm OD tube with 8mm ring
SDP
Catalog No. 50974610
Encompass_Preferred
5/16 I.D. tubing hose connection on 3mm O.D. tube, with 8mm O.D. second ring. Three ring connector is tooled with flare and exposed for direct sealing to apparatus. Approx. 5-6 between connectors.
Catalog No. 50-974-610 Supplier Ace Glass, Inc. Supplier No. 846903
5/16 I.D. tubing hose connection on 3mm O.D. tube, with 8mm O.D. second ring. Three ring connector is tooled with flare and exposed for direct sealing to apparatus. Approx. 5-6 between connectors.