Learn More
MSC Campbell Wire Rope Clip, 5/16″ Rope Dia, Steel, Electrogalvanized Finish
Catalog No. 5000534247
Encompass
This product is a wire rope clip, thimble clip, and thimble designed for a 5/16″ rope diameter. It is made of steel and serves as a rope clamp. The product has an electrogalvanized finish and requires a minimum of two clips for assembly. If used on coated cable, the coating should be removed from the assembly area.
  • Accessory type is clip, thimble clip, and thimble.
  • Designed for 5/16″ rope diameter.
  • Constructed from steel material.
  • Features a 3/8-16 thread size.
  • Requires removal of coating from assembly area if used on coated cable.
  • Minimum of two clips required for assembly.
  • Clevis attachment type.
  • Electrogalvanized finish for durability.
  • Functions as a rope clamp.
Catalog No. 50-005-34247 Supplier MSC Supplier No. 01169960
This product is a wire rope clip, thimble clip, and thimble designed for a 5/16″ rope diameter. It is made of steel and serves as a rope clamp. The product has an electrogalvanized finish and requires a minimum of two clips for assembly. If used on coated cable, the coating should be removed from the assembly area.
  • Accessory type is clip, thimble clip, and thimble.
  • Designed for 5/16″ rope diameter.
  • Constructed from steel material.
  • Features a 3/8-16 thread size.
  • Requires removal of coating from assembly area if used on coated cable.
  • Minimum of two clips required for assembly.
  • Clevis attachment type.
  • Electrogalvanized finish for durability.
  • Functions as a rope clamp.