Learn More
AVANTOR MASTERFLEX (VWR LLC) Fitting, Polypropylene, Straight, Hose Barb Union, 5/16"" ID; 10/pk
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX4061001/DEL
Encompass
Ideal for general-purpose and single-use applications, to connect tubing with rest of system.
Catalog No. NC1503230