Learn More
US Plastic Corporation 5/16" ID x 9/16" OD x 1/8" Wall Excelon RNT Clear PVC Tubing
SDP
Catalog No. NC1356526
Encompass
This clear tubing, with a dense, smooth bore, is made from the highest quality polyvinyl chloride (PVC) and special plasticizers without extenders or fillers. It is ideal for many applications in the laboratory as well as in chemical and industrial plants, and it has a minimum bend radius of 3x diameter.
Catalog No. NC1356526 Supplier US Plastic Corporation Supplier No. 59011
This clear tubing, with a dense, smooth bore, is made from the highest quality polyvinyl chloride (PVC) and special plasticizers without extenders or fillers. It is ideal for many applications in the laboratory as well as in chemical and industrial plants, and it has a minimum bend radius of 3x diameter.