Learn More
Ace Glass, Inc. PTFE 5/16in tube compression fitting to 3/8in male NPT
SDP
Catalog No. 50977680
Encompass
5/16 PTFE tube compression fitting to 3/8 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.
Catalog No. 50-977-680 Supplier Ace Glass, Inc. Supplier No. 1270934
5/16 PTFE tube compression fitting to 3/8 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.