Learn More
Medchemexpress LLC 5-[2-[(3,4-dichlorobenzoyl)amino]ethyl]-N-hydroxy-1,2-oxazole-3-carboxamide | 2245942-72-5 | 99.0% | 344.15 g·mol⁻1 | C13H11Cl2N3O4 | 1MG
SDP
Catalog No. 5000236655
Encompass_Preferred
SS-208 (AVS100) is a selective histone deacetylase 6 (HDAC6) inhibitor for research use, with a reported IC50 of 12 nM and a molecular weight of 344.15 g·mol⁻1. It has demonstrated anti-tumor activity in melanoma models and is provided as a high-purity solid for preclinical and biochemical studies.
  • Selective HDAC6 inhibition with low nanomolar potency.
  • High reported purity suitable for research applications.
  • Applicable to cell-based and biochemical assays.
  • Provided as a solid for easy handling and dissolution.
  • Includes product datasheet and safety data documentation.
Catalog No. 50-002-36655 Supplier Medchemexpress LLC Supplier No. HY1263301MG
SS-208 (AVS100) is a selective histone deacetylase 6 (HDAC6) inhibitor for research use, with a reported IC50 of 12 nM and a molecular weight of 344.15 g·mol⁻1. It has demonstrated anti-tumor activity in melanoma models and is provided as a high-purity solid for preclinical and biochemical studies.
  • Selective HDAC6 inhibition with low nanomolar potency.
  • High reported purity suitable for research applications.
  • Applicable to cell-based and biochemical assays.
  • Provided as a solid for easy handling and dissolution.
  • Includes product datasheet and safety data documentation.